Product Details

SNP ID
rs200591893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45630926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAGCAGCCTAAACTATATCTA[G/T]GCAAACACTTATGTGAATCTCACCG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WFDC10A PubMed Links

Gene Details

Gene
WFDC10A
Gene Name
WAP four-disulfide core domain 10A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080753.2 130 Intron NP_542791.1
XM_017027679.1 130 Missense Mutation GGC,TGC G44C XP_016883168.1
Gene
WFDC9
Gene Name
WAP four-disulfide core domain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147198.3 130 Intron NP_671731.1

View Full Product Details