Product Details

SNP ID
rs200345331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189563783 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGGATATCTTCAAAAGGAGAAAC[A/G]GACAAGTCCAGGGGGCTTCCAGGCA
Phenotype
MIM: 604653
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC40A1 PubMed Links

Gene Details

Gene
SLC40A1
Gene Name
solute carrier family 40 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014585.5 1689 Silent Mutation TCC,TCT S401S NP_055400.1
XM_005246505.1 1689 Silent Mutation TCC,TCT S361S XP_005246562.1
XM_017003938.1 1689 Silent Mutation TCC,TCT S361S XP_016859427.1

View Full Product Details