Product Details

SNP ID
rs202089640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113844569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGATGTAGAGGAAAATCCTAGT[C/T]CAACTGTGGAAGAGAATAATGTGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRAMD1C PubMed Links

Gene Details

Gene
GRAMD1C
Gene Name
GRAM domain containing 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172105.1 160 Intron NP_001165576.1
NM_017577.4 160 Missense Mutation CCA,TCA P32S NP_060047.3
XM_005247546.2 160 Missense Mutation CCA,TCA P32S XP_005247603.1
XM_005247547.1 160 Missense Mutation CCA,TCA P32S XP_005247604.1
XM_011512930.1 160 Missense Mutation CCA,TCA P22S XP_011511232.1
XM_011512931.1 160 Intron XP_011511233.1
XM_017006646.1 160 Missense Mutation CCA,TCA P32S XP_016862135.1
XM_017006647.1 160 UTR 5 XP_016862136.1
XM_017006648.1 160 Intron XP_016862137.1
XM_017006649.1 160 Intron XP_016862138.1

View Full Product Details