Product Details

SNP ID
rs201365038
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187371047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGAGGAAGTCTCCAGAAATGCCA[A/G]TAATCCTGGAAGTGTCCCTGGAAGG
Phenotype
MIM: 609350
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RTP4 PubMed Links

Gene Details

Gene
RTP4
Gene Name
receptor transporter protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022147.2 477 Missense Mutation ATA,GTA I139V NP_071430.2

View Full Product Details