Product Details

SNP ID
rs201557940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57073834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCCACCCAACATCCCAACAAAC[C/T]CCACTGTGCACTGGCATCATGCCAG
Phenotype
MIM: 612115 MIM: 609869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF3 PubMed Links

Gene Details

Gene
ARHGEF3
Gene Name
Rho guanine nucleotide exchange factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128615.1 1361 Intron NP_001122087.1
NM_001128616.1 1361 Intron NP_001122088.1
NM_001289698.1 1361 Intron NP_001276627.1
NM_019555.2 1361 Intron NP_062455.1
XM_005265186.4 1361 Intron XP_005265243.1
XM_005265187.3 1361 Intron XP_005265244.1
XM_011533764.1 1361 Intron XP_011532066.1
XM_011533765.1 1361 Intron XP_011532067.1
XM_011533766.2 1361 Intron XP_011532068.1
XM_011533767.1 1361 Intron XP_011532069.1
XM_017006502.1 1361 Intron XP_016861991.1
XM_017006503.1 1361 Intron XP_016861992.1
XM_017006504.1 1361 Intron XP_016861993.1
XM_017006505.1 1361 Intron XP_016861994.1
XM_017006506.1 1361 Intron XP_016861995.1
Gene
SPATA12
Gene Name
spermatogenesis associated 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181727.1 1361 Missense Mutation CCC,CTC P47L NP_859078.1
XM_011533676.2 1361 Missense Mutation CCC,CTC P47L XP_011531978.1

View Full Product Details