Product Details

SNP ID
rs200857960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:187368519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAAAACCCCGGGCCACATGGAC[A/G]CTGAAGTTGGATGGCAACCTTCAGC
Phenotype
MIM: 609350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RTP4 PubMed Links

Gene Details

Gene
RTP4
Gene Name
receptor transporter protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022147.2 140 Silent Mutation ACA,ACG T26T NP_071430.2

View Full Product Details