Product Details

SNP ID
rs199596871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48628021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGACATTAATGGAAACTGAGGC[A/G]CCCTTGGGGGAGGCAGCCTACACCA
Phenotype
MIM: 604264 MIM: 610068
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CELSR3 PubMed Links

Gene Details

Gene
CELSR3
Gene Name
cadherin EGF LAG seven-pass G-type receptor 3
There are no transcripts associated with this gene.

Gene
MIR6824
Gene Name
microRNA 6824
There are no transcripts associated with this gene.

Gene
SLC26A6
Gene Name
solute carrier family 26 member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040454.1 1918 Silent Mutation GGC,GGT G585G NP_001035544.1
NM_001281732.1 1918 Silent Mutation GGC,GGT G570G NP_001268661.1
NM_001281733.1 1918 Silent Mutation GGC,GGT G499G NP_001268662.1
NM_022911.2 1918 Silent Mutation GGC,GGT G606G NP_075062.2
NM_134263.2 1918 Silent Mutation GGC,GGT G606G NP_599025.2
NM_134426.2 1918 Intron NP_602298.2
Gene
TMEM89
Gene Name
transmembrane protein 89
There are no transcripts associated with this gene.

View Full Product Details