Product Details

SNP ID
rs199645579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196722537 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCGTATGAGTTGGCTTCATTAC[A/G]AGAGAGAAACATAACAGAGGTACAG
Phenotype
MIM: 610276
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIGX PubMed Links

Gene Details

Gene
PIGX
Gene Name
phosphatidylinositol glycan anchor biosynthesis class X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166304.1 458 Missense Mutation CAA,CGA Q100R NP_001159776.1
NM_017861.3 458 Missense Mutation CAA,CGA Q100R NP_060331.3

View Full Product Details