Product Details

SNP ID
rs200718093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70335115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTAAAACACCAACTGCAGCAACC[A/G]TTTTCTTTGGGGCTGACAATGCTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CABS1 PubMed Links

Gene Details

Gene
CABS1
Gene Name
calcium binding protein, spermatid associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033122.3 162 Missense Mutation ATT,GTT I26V NP_149113.3

View Full Product Details