Product Details

SNP ID
rs201448780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:70335649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATCCAATGTCCCTGCTGATGAG[G/T]CTGTCCAGGTCACTGATTCCACTAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CABS1 PubMed Links

Gene Details

Gene
CABS1
Gene Name
calcium binding protein, spermatid associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033122.3 696 Missense Mutation GCT,TCT A204S NP_149113.3

View Full Product Details