Product Details

SNP ID
rs201621075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55952239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTTACCTTTGGTAGAAAAACTT[C/T]TCAGGTAAAGACAAAAATACAGTTT
Phenotype
MIM: 611423
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP135 PubMed Links

Gene Details

Gene
CEP135
Gene Name
centrosomal protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025009.4 377 Missense Mutation CTC,TTC L37F NP_079285.2
XM_005265788.3 377 UTR 5 XP_005265845.1
XM_006714055.2 377 Missense Mutation CTC,TTC L37F XP_006714118.1
XM_011534412.2 377 Intron XP_011532714.1

View Full Product Details