Product Details

SNP ID
rs201690440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:56338733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGAACCAAAAAATATTTTTTGC[C/T]CTGATGGTGAGGTACACGGGGATGA
Phenotype
MIM: 614365
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AASDH PubMed Links

Gene Details

Gene
AASDH
Gene Name
aminoadipate-semialdehyde dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286668.1 2931 Missense Mutation GAG,GGG E889G NP_001273597.1
NM_001286669.1 2931 Missense Mutation GAG,GGG E836G NP_001273598.1
NM_001286670.1 2931 Missense Mutation GAG,GGG E504G NP_001273599.1
NM_001286671.1 2931 Silent Mutation AGA,AGG R849R NP_001273600.1
NM_001286672.1 2931 Intron NP_001273601.1
NM_001323890.1 2931 Missense Mutation GAG,GGG E1028G NP_001310819.1
NM_001323892.1 2931 Silent Mutation AGA,AGG R934R NP_001310821.1
NM_001323893.1 2931 Silent Mutation AGA,AGG R781R NP_001310822.1
NM_001323899.1 2931 Missense Mutation GAG,GGG E612G NP_001310828.1
NM_181806.3 2931 Missense Mutation GAG,GGG E989G NP_861522.2
XM_017007740.1 2931 Missense Mutation GAG,GGG E945G XP_016863229.1
XM_017007741.1 2931 Missense Mutation GAG,GGG E836G XP_016863230.1
XM_017007742.1 2931 Missense Mutation GAG,GGG E836G XP_016863231.1
XM_017007743.1 2931 Silent Mutation AGA,AGG R890R XP_016863232.1
XM_017007744.1 2931 Missense Mutation GAG,GGG E567G XP_016863233.1
Gene
KIAA1211
Gene Name
KIAA1211
There are no transcripts associated with this gene.

View Full Product Details