Product Details

SNP ID
rs201675155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140645634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCAGGGCTCTGGTTACCTGAT[C/T]AGCACTGAAGTTGTTCAAAGGGACA
Phenotype
MIM: 600549 MIM: 602137
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IK PubMed Links

Gene Details

Gene
IK
Gene Name
IK cytokine, down-regulator of HLA II
There are no transcripts associated with this gene.

Gene
MIR3655
Gene Name
microRNA 3655
There are no transcripts associated with this gene.

Gene
NDUFA2
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185012.1 455 UTR 3 NP_001171941.1
NM_002488.4 455 Missense Mutation AAT,GAT N85D NP_002479.1
Gene
TMCO6
Gene Name
transmembrane and coiled-coil domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300980.1 455 Intron NP_001287909.1
NM_001300982.1 455 Intron NP_001287911.1
NM_018502.4 455 Intron NP_060972.3
XM_005268477.1 455 Intron XP_005268534.1
XM_011537663.2 455 Intron XP_011535965.1
XM_011537665.2 455 Intron XP_011535967.1
XM_011537668.2 455 Intron XP_011535970.1
XM_017009617.1 455 Intron XP_016865106.1
XM_017009618.1 455 Intron XP_016865107.1
XM_017009619.1 455 Intron XP_016865108.1
XM_017009620.1 455 Intron XP_016865109.1

View Full Product Details