Product Details

SNP ID
rs201808800
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35805703 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGCTGCCGGCCCAGGAGGCAGC[A/C]AAGATCTACCATACCAACTATGTGC
Phenotype
MIM: 120105 MIM: 609427
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLPS PubMed Links

Gene Details

Gene
CLPS
Gene Name
colipase
There are no transcripts associated with this gene.

Gene
LHFPL5
Gene Name
lipoma HMGIC fusion partner-like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182548.3 410 Silent Mutation GCA,GCC A11A NP_872354.1

View Full Product Details