Product Details

SNP ID
rs200781115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99621409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCAGGGAGAGCAGGAGGAAACA[A/G]CACTTTGCAGAGGCGCTTGGGAGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN25 PubMed Links

Gene Details

Gene
ZSCAN25
Gene Name
zinc finger and SCAN domain containing 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145115.2 1006 Missense Mutation ACA,GCA T142A NP_660090.2
XM_005250194.2 1006 Missense Mutation ACA,GCA T142A XP_005250251.1
XM_011515905.2 1006 Missense Mutation ACA,GCA T142A XP_011514207.1
XM_011515906.2 1006 Missense Mutation ACA,GCA T142A XP_011514208.1
XM_011515907.2 1006 Missense Mutation ACA,GCA T142A XP_011514209.1
XM_011515908.2 1006 UTR 5 XP_011514210.1
XM_011515909.2 1006 Missense Mutation ACA,GCA T142A XP_011514211.1
XM_011515910.2 1006 Missense Mutation ACA,GCA T142A XP_011514212.1
XM_017011824.1 1006 Missense Mutation ACA,GCA T142A XP_016867313.1
XM_017011825.1 1006 UTR 5 XP_016867314.1
XM_017011826.1 1006 UTR 5 XP_016867315.1
XM_017011827.1 1006 Missense Mutation ACA,GCA T142A XP_016867316.1

View Full Product Details