Product Details

SNP ID
rs201194192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:20785097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGCAGCGCAGCCGCGCTGTTCGG[C/G]TTCTGCACGTCGATCCAGCCGGCCC
Phenotype
MIM: 608306
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SP8 PubMed Links

Gene Details

Gene
SP8
Gene Name
Sp8 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182700.5 907 Missense Mutation AAC,AAG N240K NP_874359.2
NM_198956.3 907 Missense Mutation AAC,AAG N222K NP_945194.1

View Full Product Details