Product Details

SNP ID
rs199903761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:9556111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGCGGCCTGGCCCCCTTCGCC[C/T]CCCCGCGGCACGGCCTAGCGCTGCC
Phenotype
MIM: 603303
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNKS PubMed Links

Gene Details

Gene
TNKS
Gene Name
tankyrase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003747.2 883 Missense Mutation CCC,TCC P58S NP_003738.2
XM_006716263.3 883 Missense Mutation CCC,TCC P58S XP_006716326.1
XM_011543845.2 883 Missense Mutation CCC,TCC P58S XP_011542147.1
XM_011543846.2 883 Missense Mutation CCC,TCC P58S XP_011542148.1
XM_011543847.2 883 Missense Mutation CCC,TCC P58S XP_011542149.1

View Full Product Details