Product Details
- SNP ID
-
rs202137559
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:98808057 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTGCACCGCCGCCTGCCCGAGC[C/G]CTGGAACCCGCTGTGAGTGCACAGC
- Phenotype
-
MIM: 610290
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GALNT12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1137654] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GALNT12
- Gene Name
- polypeptide N-acetylgalactosaminyltransferase 12
View Full Product Details