Product Details

SNP ID
rs201644989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137255954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTCTTCAGTCAGCCCTCCCCTT[C/T]TTGGACCTGCACGGGACGCCGCGGC
Phenotype
MIM: 611180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf173-AS1 PubMed Links

Gene Details

Gene
C9orf173-AS1
Gene Name
C9orf173 antisense RNA 1
There are no transcripts associated with this gene.

Gene
FAM166A
Gene Name
family with sequence similarity 166 member A
There are no transcripts associated with this gene.

Gene
NELFB
Gene Name
negative elongation factor complex member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015456.4 360 Silent Mutation TTC,TTT F98F NP_056271.3
Gene
STPG3
Gene Name
sperm-tail PG-rich repeat containing 3
There are no transcripts associated with this gene.

View Full Product Details