Product Details

SNP ID
rs555070491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:63061275 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCATTGAATGCTCTCAGAAATGC[T/G]GCTGAGATAATCTTCAGACTTCATT
Phenotype
MIM: 600415
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
TTPA PubMed Links

Gene Details

Gene
TTPA
Gene Name
alpha tocopherol transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000370.3 968 Missense Mutation AGC,CGC S272R NP_000361.1
XM_006716468.3 968 Missense Mutation AGC,CGC S156R XP_006716531.1

View Full Product Details