Product Details

SNP ID
rs367862706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17393090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGATGGCACCCCGCTTCAGGAC[G/A]ATCACCAGGTCTGCACTCAGGATGG
Phenotype
MIM: 600509 MIM: 600937
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
ABCC8 PubMed Links

Gene Details

Gene
ABCC8
Gene Name
ATP binding cassette subfamily C member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000352.4 4776 Silent Mutation ATC,ATT I1549I NP_000343.2
NM_001287174.1 4776 Silent Mutation ATC,ATT I1550I NP_001274103.1
XM_011520331.2 4776 Silent Mutation ATC,ATT I1549I XP_011518633.1
XM_017018197.1 4776 Silent Mutation ATC,ATT I1572I XP_016873686.1
XM_017018198.1 4776 Silent Mutation ATC,ATT I1571I XP_016873687.1
XM_017018199.1 4776 Silent Mutation ATC,ATT I1571I XP_016873688.1
XM_017018200.1 4776 Silent Mutation ATC,ATT I1548I XP_016873689.1
XM_017018201.1 4776 Intron XP_016873690.1
XM_017018202.1 4776 Silent Mutation ATC,ATT I1071I XP_016873691.1
XM_017018203.1 4776 Intron XP_016873692.1
XM_017018204.1 4776 Silent Mutation ATC,ATT I868I XP_016873693.1
Gene
KCNJ11
Gene Name
potassium voltage-gated channel subfamily J member 11
There are no transcripts associated with this gene.

View Full Product Details