Product Details

SNP ID
rs532620482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88427927 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCAGCTCCCTGAGGTGGACACC[C/G]CCCAGGGCCCTGGGACTGGAGCTCC
Phenotype
MIM: 612078
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF469 PubMed Links

Gene Details

Gene
ZNF469
Gene Name
zinc finger protein 469
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127464.2 650 Missense Mutation CCC,GCC P153A NP_001120936.2
XM_017023784.1 650 Missense Mutation CCC,GCC P153A XP_016879273.1
XM_017023785.1 650 Missense Mutation CCC,GCC P153A XP_016879274.1

View Full Product Details