Product Details

SNP ID
rs568661500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88427453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCGGACAGCTGCGTCGTCCTAG[C/A]GCCAGGACGGAGGGGCCATGCCTGG
Phenotype
MIM: 612078
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
ZNF469 PubMed Links

Gene Details

Gene
ZNF469
Gene Name
zinc finger protein 469
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127464.2 176 Intron NP_001120936.2
XM_017023784.1 176 UTR 5 XP_016879273.1
XM_017023785.1 176 UTR 5 XP_016879274.1

View Full Product Details