Product Details
- SNP ID
-
rs866626696
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:10638857 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTTTAAGGCTCTTGATTATTAA[G/T]TTAATAAATCAAATATACACAGTGA
- Phenotype
-
MIM: 601920
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
JAG1
PubMed Links
Gene Details
- Gene
- JAG1
- Gene Name
- jagged 1
View Full Product Details