Product Details

SNP ID
rs760670617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:118723570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACAACAAAGGAAATCCACATAA[G/C]AAGAGCCTCCGTAAAGTCCTTTGTC
Phenotype
MIM: 607186
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
SEC24D PubMed Links

Gene Details

Gene
SEC24D
Gene Name
SEC24 homolog D, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318066.1 2499 Missense Mutation TCT,TGT S1016C NP_001304995.1
NM_014822.3 2499 Missense Mutation TCT,TGT S1015C NP_055637.2
XM_005263379.2 2499 Missense Mutation TCT,TGT S1016C XP_005263436.1
XM_017008875.1 2499 Missense Mutation TCT,TGT S571C XP_016864364.1

View Full Product Details