Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001282116.1 | 6725 | UTR 3 | NP_001269045.1 | ||
NM_001282117.1 | 6725 | Intron | NP_001269046.1 | ||
NM_002919.3 | 6725 | Intron | NP_002910.1 | ||
NM_134428.2 | 6725 | UTR 3 | NP_602304.1 | ||
XM_006716840.1 | 6725 | UTR 3 | XP_006716903.1 | ||
XM_006716843.1 | 6725 | UTR 3 | XP_006716906.1 | ||
XM_006716844.1 | 6725 | UTR 3 | XP_006716907.1 | ||
XM_006716845.2 | 6725 | Intron | XP_006716908.1 | ||
XM_006716847.3 | 6725 | UTR 3 | XP_006716910.1 | ||
XM_011517998.1 | 6725 | UTR 3 | XP_011516300.1 | ||
XM_011517999.1 | 6725 | UTR 3 | XP_011516301.1 | ||
XM_011518000.1 | 6725 | UTR 3 | XP_011516302.1 | ||
XM_011518001.1 | 6725 | UTR 3 | XP_011516303.1 | ||
XM_017015000.1 | 6725 | UTR 3 | XP_016870489.1 | ||
XM_017015001.1 | 6725 | UTR 3 | XP_016870490.1 | ||
XM_017015002.1 | 6725 | UTR 3 | XP_016870491.1 |