Product Details

SNP ID
rs3924257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139418036 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACAGAGGAAGATGTGGGGCAGG[A/C]GGGTGTTTGAAGAATGTGAAATGCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC18 PubMed Links

Gene Details

Gene
DNAJC18
Gene Name
DnaJ heat shock protein family (Hsp40) member C18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152686.3 Intron NP_689899.1

View Full Product Details