Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001317934.1 | 286 | Intron | NP_001304863.1 | ||
NM_001317935.1 | 286 | Intron | NP_001304864.1 | ||
NM_020894.3 | 286 | Intron | NP_065945.2 | ||
XM_017008490.1 | 286 | Intron | XP_016863979.1 | ||
XM_017008491.1 | 286 | Intron | XP_016863980.1 | ||
XM_017008492.1 | 286 | Intron | XP_016863981.1 | ||
XM_017008493.1 | 286 | Intron | XP_016863982.1 | ||
XM_017008494.1 | 286 | Intron | XP_016863983.1 | ||
XM_017008495.1 | 286 | Intron | XP_016863984.1 | ||
XM_017008496.1 | 286 | UTR 5 | XP_016863985.1 | ||
XM_017008497.1 | 286 | UTR 5 | XP_016863986.1 | ||
XM_017008498.1 | 286 | Intron | XP_016863987.1 | ||
XM_017008499.1 | 286 | Intron | XP_016863988.1 | ||
XM_017008500.1 | 286 | Intron | XP_016863989.1 |