Product Details
- SNP ID
-
rs756924
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:2301799 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CGTCTCCTCCACATCCCACACCAGG[C/T]CCTGCCCATGACTTTCCCCCTCTCA
- Phenotype
-
MIM: 611033
MIM: 603853
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C11orf21
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114342362] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C11orf21
- Gene Name
- chromosome 11 open reading frame 21
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
XM_011520034.1 |
115 |
Silent Mutation |
GGA,GGG |
G21G |
XP_011518336.1 |
- Gene
- TSPAN32
- Gene Name
- tetraspanin 32
View Full Product Details