Product Details

SNP ID
rs710193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40827186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCATGCCCCCTCCCATTCCAGG[C/T]ATTCCTCCGGGAAAATTACCAGGCA
Phenotype
MIM: 606795 MIM: 606796
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A17 PubMed Links

Gene Details

Gene
SLC25A17
Gene Name
solute carrier family 25 member 17
There are no transcripts associated with this gene.

Gene
ST13
Gene Name
suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278589.1 1359 Missense Mutation ATA,ATG I287M NP_001265518.1
NM_003932.4 1359 Missense Mutation ATA,ATG I297M NP_003923.2

View Full Product Details