Product Details

SNP ID
rs11673314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43476359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCATTCTACACGTGAGAAAACTCA[A/G]GCCGGCCGGGCTGGGTGGCTCACGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHLDB3 PubMed Links

Gene Details

Gene
PHLDB3
Gene Name
pleckstrin homology like domain family B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198850.3 Intron NP_942147.3
XM_005259169.3 Intron XP_005259226.1
XM_005259170.3 Intron XP_005259227.1
XM_005259171.3 Intron XP_005259228.1
XM_011527236.2 Intron XP_011525538.1

View Full Product Details