Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130079.2 | 4766 | Intron | NP_001123551.1 | ||
NM_001286072.1 | 4766 | Intron | NP_001273001.1 | ||
NM_001321324.1 | 4766 | Intron | NP_001308253.1 | ||
NM_020963.4 | 4766 | Intron | NP_066014.1 | ||
XM_005270869.4 | 4766 | UTR 3 | XP_005270926.1 | ||
XM_005270870.4 | 4766 | UTR 3 | XP_005270927.1 | ||
XM_017001318.1 | 4766 | UTR 3 | XP_016856807.1 | ||
XM_017001319.1 | 4766 | Intron | XP_016856808.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001042678.1 | 4766 | Intron | NP_001036143.1 | ||
NM_001042679.1 | 4766 | Intron | NP_001036144.1 | ||
NM_175744.4 | 4766 | Intron | NP_786886.1 |