Product Details

SNP ID
rs12974195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48277744 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGGGTGTTCATTGCCTCCTGCTT[C/G]TTAATTCTTCTAAGCATTTTCTATG
Phenotype
MIM: 603996
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF114 PubMed Links

Gene Details

Gene
ZNF114
Gene Name
zinc finger protein 114
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301062.1 Intron NP_001287991.1
NM_153608.2 Intron NP_705836.1
XM_011526574.2 Intron XP_011524876.1
XM_017026414.1 Intron XP_016881903.1
XM_017026415.1 Intron XP_016881904.1

View Full Product Details