Product Details
- SNP ID
-
rs2042404
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:58472430 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTAGAGCATGGTGTTGCCATTCGA[A/G]TGTGGTGGGAAAGGCCTGCTGTCAA
- Phenotype
-
MIM: 614463
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NDRG4
PubMed Links
Gene Details
- Gene
- NDRG4
- Gene Name
- NDRG family member 4
View Full Product Details