Product Details

SNP ID
rs1568673
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:151833734 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAAAATATGACTGCCAAATTTAAC[A/G]GTGGATTTTGTTATAAGTTTGTGCT
Phenotype
MIM: 608960 MIM: 601949
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL5A PubMed Links

Gene Details

Gene
ARL5A
Gene Name
ADP ribosylation factor like GTPase 5A
There are no transcripts associated with this gene.

Gene
CACNB4
Gene Name
calcium voltage-gated channel auxiliary subunit beta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011511795.2 7736 UTR 3 XP_011510097.1
XM_011511796.2 7736 UTR 3 XP_011510098.1
XM_011511797.2 7736 UTR 3 XP_011510099.1
XM_011511798.2 7736 Intron XP_011510100.1
XM_017004880.1 7736 UTR 3 XP_016860369.1
XM_017004881.1 7736 UTR 3 XP_016860370.1
XM_017004882.1 7736 UTR 3 XP_016860371.1
XM_017004883.1 7736 UTR 3 XP_016860372.1
XM_017004884.1 7736 UTR 3 XP_016860373.1
XM_017004885.1 7736 UTR 3 XP_016860374.1
XM_017004886.1 7736 UTR 3 XP_016860375.1
XM_017004887.1 7736 Intron XP_016860376.1
XM_017004888.1 7736 Intron XP_016860377.1

View Full Product Details