Product Details

SNP ID
rs8039119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89691880 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTATAAATCAAGCAACAAATTGT[A/T]AAGTAAAATATAGTCTGTTTTCCTA
Phenotype
MIM: 603866
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PEX11A PubMed Links

Gene Details

Gene
PEX11A
Gene Name
peroxisomal biogenesis factor 11 alpha
There are no transcripts associated with this gene.

Gene
WDR93
Gene Name
WD repeat domain 93
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284395.1 Intron NP_001271324.1
NM_001284396.1 Intron NP_001271325.1
NM_020212.1 Intron NP_064597.1
XM_006720614.1 Intron XP_006720677.1
XM_006720615.1 Intron XP_006720678.1
XM_006720618.2 Intron XP_006720681.1
XM_011521794.2 Intron XP_011520096.1
XM_011521795.1 Intron XP_011520097.1
XM_011521796.2 Intron XP_011520098.1
XM_011521798.2 Intron XP_011520100.1
XM_011521800.1 Intron XP_011520102.1
XM_011521802.2 Intron XP_011520104.1
XM_017022415.1 Intron XP_016877904.1
XM_017022416.1 Intron XP_016877905.1
XM_017022417.1 Intron XP_016877906.1

View Full Product Details