Product Details

SNP ID
rs80359828
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:42927173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTTTAGTCTCAGGAACTTTGAA[G/T]TAGGTGAAGATGAAGAACAGAACCA
Phenotype
MIM: 138140 MIM: 143090
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC2A1 PubMed Links

Gene Details

Gene
SLC2A1
Gene Name
solute carrier family 2 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006516.2 1872 Nonsense Mutation TAA,TAC *449Y NP_006507.2

View Full Product Details