Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_003607.3 | 9428 | UTR 3 | NP_003598.2 | ||
NM_014826.4 | 9428 | UTR 3 | NP_055641.3 | ||
XM_005273317.3 | 9428 | Intron | XP_005273374.1 | ||
XM_005273318.3 | 9428 | Intron | XP_005273375.1 | ||
XM_005273320.3 | 9428 | Intron | XP_005273377.1 | ||
XM_005273321.3 | 9428 | UTR 3 | XP_005273378.1 | ||
XM_005273322.3 | 9428 | UTR 3 | XP_005273379.1 | ||
XM_005273324.3 | 9428 | UTR 3 | XP_005273381.1 | ||
XM_006711834.3 | 9428 | Intron | XP_006711897.1 | ||
XM_006711835.3 | 9428 | Intron | XP_006711898.1 | ||
XM_011544306.2 | 9428 | Intron | XP_011542608.1 | ||
XM_011544307.2 | 9428 | Intron | XP_011542609.1 | ||
XM_011544308.2 | 9428 | UTR 3 | XP_011542610.1 | ||
XM_011544309.2 | 9428 | Intron | XP_011542611.1 | ||
XM_017002578.1 | 9428 | UTR 3 | XP_016858067.1 | ||
XM_017002579.1 | 9428 | Intron | XP_016858068.1 | ||
XM_017002580.1 | 9428 | Intron | XP_016858069.1 | ||
XM_017002581.1 | 9428 | Intron | XP_016858070.1 | ||
XM_017002582.1 | 9428 | Intron | XP_016858071.1 |