Product Details

SNP ID
rs3027506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48935070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGTCAGAGATGCCAGAGAAGAG[G/C]TCTGAATCATAGAACATCCTTTGGG
Phenotype
MIM: 300713
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
OTUD5 PubMed Links

Gene Details

Gene
OTUD5
Gene Name
OTU deubiquitinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136157.1 Intron NP_001129629.1
NM_001136158.1 Intron NP_001129630.1
NM_001136159.1 Intron NP_001129631.1
NM_017602.3 Intron NP_060072.1
XM_005272622.3 Intron XP_005272679.1
XM_005278039.3 Intron XP_005278096.1
XM_005278042.4 Intron XP_005278099.1
XM_006724537.2 Intron XP_006724600.1
XM_011543932.1 Intron XP_011542234.1
XM_011543934.2 Intron XP_011542236.1
XM_017029628.1 Intron XP_016885117.1
XM_017029629.1 Intron XP_016885118.1
XM_017029630.1 Intron XP_016885119.1

View Full Product Details