Product Details

SNP ID
rs2374256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90214055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACATTTTATGACGATTTACATTT[A/T]AAGATGATTTTAGCTGCTGTGGAGA
Phenotype
MIM: 605094
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STEAP2 PubMed Links

Gene Details

Gene
STEAP2
Gene Name
STEAP2 metalloreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040665.1 1587 Intron NP_001035755.1
NM_001040666.1 1587 Intron NP_001035756.1
NM_001244944.1 1587 Intron NP_001231873.1
NM_001244945.1 1587 Intron NP_001231874.1
NM_001244946.1 1587 Intron NP_001231875.1
NM_152999.3 1587 Intron NP_694544.2
XM_006715921.3 1587 Intron XP_006715984.1
XM_017011953.1 1587 Intron XP_016867442.1
XM_017011954.1 1587 Intron XP_016867443.1
XM_017011955.1 1587 Intron XP_016867444.1
XM_017011956.1 1587 Intron XP_016867445.1
XM_017011957.1 1587 Intron XP_016867446.1
XM_017011958.1 1587 UTR 5 XP_016867447.1
Gene
STEAP2-AS1
Gene Name
STEAP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details