Product Details

SNP ID
rs2186616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:86795486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCACATCCTTTGCTACTCCGGGAT[C/G]CCATGGTAGGCACCGTGGGTGCCCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRSS23 PubMed Links

Gene Details

Gene
PRSS23
Gene Name
protease, serine 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293178.1 Intron NP_001280107.1
NM_001293179.1 Intron NP_001280108.1
NM_001293180.1 Intron NP_001280109.1
NM_007173.5 Intron NP_009104.2

View Full Product Details