Product Details

SNP ID
rs2229653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29663793 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCCAGACGCTCTGGGGAGCACAA[T/C]AGCAGTGCAGACACCCACCTCCGGA
Phenotype
MIM: 606248 MIM: 182160
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
QPRT PubMed Links

Gene Details

Gene
QPRT
Gene Name
quinolinate phosphoribosyltransferase
There are no transcripts associated with this gene.

Gene
SPN
Gene Name
sialophorin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030288.2 230 Missense Mutation ACA,ATA T22I NP_001025459.1
NM_003123.4 230 Missense Mutation ACA,ATA T22I NP_003114.1

View Full Product Details