Product Details

SNP ID
rs2231180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:904710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTTTCTGACTTTCCCTCTTTC[C/T]CTTAATCGCCTGCTTCTTCCTCCGG
Phenotype
MIM: 102980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADCYAP1 PubMed Links

Gene Details

Gene
ADCYAP1
Gene Name
adenylate cyclase activating polypeptide 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099733.1 Intron NP_001093203.1
NM_001117.4 Intron NP_001108.2
XM_005258081.3 Intron XP_005258138.1

View Full Product Details