Product Details

SNP ID
rs2069208
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:85651900 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGTAATTCACTAAAACACACGTG[C/T]ATTGTAAATCTTACGGGAAAAAATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNHIT6 PubMed Links

Gene Details

Gene
ZNHIT6
Gene Name
zinc finger HIT-type containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170670.1 3720 UTR 3 NP_001164141.1
NM_017953.3 3720 UTR 3 NP_060423.3
XM_011541614.2 3720 Intron XP_011539916.1

View Full Product Details