Product Details
- SNP ID
-
rs2069774
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:122451160 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGTGAGCCTCCTCACAGCACTGGG[C/T]AGGGTGTAAGATATTGGAGAATTCA
- Phenotype
-
MIM: 147680
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL2
PubMed Links
Gene Details
- Gene
- IL2
- Gene Name
- interleukin 2
View Full Product Details