Product Details

SNP ID
rs2275729
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:92692673 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGGCACGTCCCGACGCGGGCTCGT[G/T]GCAAGTTTTCTTTCTCTCTTTCCTG
Phenotype
MIM: 604420
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HHEX PubMed Links

Gene Details

Gene
HHEX
Gene Name
hematopoietically expressed homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002729.4 Intron NP_002720.1

View Full Product Details