Product Details

SNP ID
rs2232947
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62242319 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAACAAGCCACCATGAAGCTGTC[G/T]GTGTGTCTCCTGCTGGTCACGCTGG
Phenotype
MIM: 615061
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SCGB1D2 PubMed Links

Gene Details

Gene
SCGB1D2
Gene Name
secretoglobin family 1D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006551.3 68 Silent Mutation TCG,TCT S4S NP_006542.1

View Full Product Details