Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001142531.1 | Intron | NP_001136003.1 | |||
NM_001142532.1 | Intron | NP_001136004.1 | |||
NM_001142533.1 | Intron | NP_001136005.1 | |||
NM_001142534.1 | Intron | NP_001136006.1 | |||
NM_001252334.1 | Intron | NP_001239263.1 | |||
NM_005489.3 | Intron | NP_005480.2 | |||
NM_170600.2 | Intron | NP_733745.1 | |||
XM_005251639.1 | Intron | XP_005251696.1 | |||
XM_011518114.1 | Intron | XP_011516416.1 | |||
XM_011518115.1 | Intron | XP_011516417.1 | |||
XM_011518117.2 | Intron | XP_011516419.1 | |||
XM_017014174.1 | Intron | XP_016869663.1 |