Product Details

SNP ID
rs2233795
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10018879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGGAGGCGGCAGGGCCACAT[C/T]GTGGTGATCAGCAGTGTCATGGGCC
Phenotype
MIM: 120216 MIM: 608575
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL5A3 PubMed Links

Gene Details

Gene
COL5A3
Gene Name
collagen type V alpha 3
There are no transcripts associated with this gene.

Gene
RDH8
Gene Name
retinol dehydrogenase 8 (all-trans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015725.2 660 Silent Mutation ATC,ATT I157I NP_056540.2

View Full Product Details