Product Details
- SNP ID
-
rs2235840
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:102515511 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGATAGAAGCACAAGAGCAGACAGG[G/A]ACAGAATAAAAAATAAAGACTGGCT
- Phenotype
-
MIM: 300939
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMSB15A
PubMed Links
Gene Details
- Gene
- TMSB15A
- Gene Name
- thymosin beta 15a
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021992.2 |
|
Intron |
|
|
NP_068832.1 |
View Full Product Details